siRNA Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022382.1 | NP_071777.1 | 25 | 3754 | ||
| GenBank mRNA | AF139185.1 | - | 25 | 3754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde4dip| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022382.1 | NP_071777.1 | 25 | 3754 | ||
| GenBank mRNA | AF139185.1 | - | 25 | 3754 | ||