siRNA Details
| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022394.1 | NP_071789.1 | 16, 17 | 2469 | ||
| GenBank mRNA | AF056324.1 | - | 16, 17 | 2469 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Safb| Gene Symbol: Safb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022394.1 | NP_071789.1 | 16, 17 | 2469 | ||
| GenBank mRNA | AF056324.1 | - | 16, 17 | 2469 | ||