siRNA Details
| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022395.2 | NP_071790.1 | 8 | 911 | ||
| GenBank mRNA | BC078826.1 | - | 8 | 906 | 7113892 | |
| D13907.1 | - | 8 | 889 | |||
| L12965.1 | - | 8 | 912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmpcb| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022395.2 | NP_071790.1 | 8 | 911 | ||
| GenBank mRNA | BC078826.1 | - | 8 | 906 | 7113892 | |
| D13907.1 | - | 8 | 889 | |||
| L12965.1 | - | 8 | 912 | |||