siRNA Details
| Gene Symbol: Calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022399.2 | NP_071794.1 | 2, 3 | 259 | ||
| GenBank mRNA | BC062395.1 | - | 2, 3 | 236 | 5623178 | |
| BU671467.1 | - | 2, 3 | 259 | |||
| D78308.1 | - | 2, 3 | 211 | |||
| FQ210587.1 | - | 264 | ||||
| FQ220096.1 | - | 2, 3 | 265 | |||
| X53363.1 | - | 2, 3 | 231 | |||
| X79327.1 | - | 2, 3 | 209 | |||

Calr
Genomic Map