siRNA Details
| Gene Symbol: Calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022399.2 | NP_071794.1 | 6 | 792 | ||
| GenBank mRNA | BC062395.1 | - | 6 | 769 | 5623178 | |
| D78308.1 | - | 6 | 744 | |||
| FQ210587.1 | - | 797 | ||||
| FQ220096.1 | - | 6 | 798 | |||
| X53363.1 | - | 6 | 764 | |||
| X79327.1 | - | 6 | 742 | |||

Calr
Genomic Map