siRNA Details
| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022512.2 | NP_071957.1 | 2 | 95 | ||
| GenBank mRNA | BC072545.1 | - | 2 | 114 | 7105799 | |
| J05030.1 | - | 2 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acads| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022512.2 | NP_071957.1 | 2 | 95 | ||
| GenBank mRNA | BC072545.1 | - | 2 | 114 | 7105799 | |
| J05030.1 | - | 2 | 96 | |||