siRNA Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022522.2 | NP_071967.2 | 11 | 1252 | ||
| XM_006236394.2 | XP_006236456.1 | 10 | 1933 | |||
| GenBank mRNA | AF136231.1 | - | 11 | 1250 | ||
| AF136232.1 | - | 12 | 1311 | |||
| U77933.1 | - | 11 | 1252 | |||

Casp2
Genomic Map