siRNA Details
| Gene Symbol: Srpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022524.1 | NP_071969.1 | 10 | 1303 | ||
| XM_006256683.2 | XP_006256745.1 | 9 | 1331 | |||
| XM_008773077.1 | XP_008771299.1 | 9 | 2457 | |||
| GenBank mRNA | BC087639.1 | - | 9 | 1302 | 7309360 | |
| D78359.1 | - | 10 | 1303 | |||

Srpx
Genomic Map