siRNA Details
| Gene Symbol: Cd200r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023953.1 | NP_076443.1 | 4 | 496 | ||
| XM_006248347.2 | XP_006248409.1 | 4 | 809 | |||
| XM_008768743.1 | XP_008766965.1 | 4 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd200r1| Gene Symbol: Cd200r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023953.1 | NP_076443.1 | 4 | 496 | ||
| XM_006248347.2 | XP_006248409.1 | 4 | 809 | |||
| XM_008768743.1 | XP_008766965.1 | 4 | 796 | |||