siRNA Details
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022534.1 | NP_071979.1 | 7 | 1119 | ||
GenBank mRNA | AF054810.1 | - | 7 | 1119 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022534.1 | NP_071979.1 | 7 | 1119 | ||
GenBank mRNA | AF054810.1 | - | 7 | 1119 |