siRNA Details
| Gene Symbol: Prpsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022545.3 | NP_071990.2 | 1, 2 | 334 | ||
| GenBank mRNA | BC078822.1 | - | 1, 2 | 309 | 7128451 | |
| D26073.1 | - | 1, 2 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpsap1| Gene Symbol: Prpsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022545.3 | NP_071990.2 | 1, 2 | 334 | ||
| GenBank mRNA | BC078822.1 | - | 1, 2 | 309 | 7128451 | |
| D26073.1 | - | 1, 2 | 232 | |||