siRNA Details
| Gene Symbol: Prpsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022545.3 | NP_071990.2 | 3 | 416 | ||
| XM_008768391.1 | XP_008766613.1 | 3 | 353 | |||
| GenBank mRNA | BC078822.1 | - | 3 | 391 | 7128451 | |
| D26073.1 | - | 3 | 314 | |||

Prpsap1
Genomic Map