siRNA Details
| Gene Symbol: Syt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031693.2 | NP_113881.1 | 2 | 789 | ||
| GenBank mRNA | DQ181551.1 | - | 2 | 553 | ||
| FQ211592.1 | - | 2 | 786 | |||
| L38247.1 | - | 2 | 790 | |||
| U14398.1 | - | 2 | 818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt4| Gene Symbol: Syt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031693.2 | NP_113881.1 | 2 | 789 | ||
| GenBank mRNA | DQ181551.1 | - | 2 | 553 | ||
| FQ211592.1 | - | 2 | 786 | |||
| L38247.1 | - | 2 | 790 | |||
| U14398.1 | - | 2 | 818 | |||