siRNA Details
| Gene Symbol: Rasd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270954.1 | NP_001257883.1 | 2 | 513 | ||
| XM_008767871.1 | XP_008766093.1 | 3 | 2465 | |||
| XM_008767872.1 | XP_008766094.1 | 3 | 2469 | |||
| XM_008767873.1 | XP_008766095.1 | 4 | 5118 | |||
| GenBank mRNA | AF239157.1 | - | 2 | 541 | ||
| BC099136.1 | - | 2 | 513 | 7455971 | ||

Rasd1
Genomic Map