siRNA Details
| Gene Symbol: Spon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172067.1 | NP_742064.1 | 11, 12 | 1718 | ||
| GenBank mRNA | BC107655.1 | - | 11, 12 | 1795 | 7935103 | |
| M88469.1 | - | 11, 12 | 1718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spon1| Gene Symbol: Spon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172067.1 | NP_742064.1 | 11, 12 | 1718 | ||
| GenBank mRNA | BC107655.1 | - | 11, 12 | 1795 | 7935103 | |
| M88469.1 | - | 11, 12 | 1718 | |||