siRNA Details
| Gene Symbol: Masp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172043.1 | NP_742040.1 | 7 | 957 | ||
| XM_008764285.1 | XP_008762507.1 | 7 | 1013 | |||
| GenBank mRNA | AJ277747.1 | - | 6 | 899 | ||
| BC128700.1 | - | 7 | 958 | 7324989 | ||
| Y18564.1 | - | 6 | 736 | |||
| Y18566.1 | - | 4 | 489 | |||
| Y18572.1 | - | 6 | 794 | |||

Masp2
Genomic Map