siRNA Details
| Gene Symbol: Masp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172043.1 | NP_742040.1 | 8, 9 | 1115 | ||
| GenBank mRNA | AJ277747.1 | - | 7, 8 | 1057 | ||
| BC128700.1 | - | 8, 9 | 1116 | 7324989 | ||
| Y18564.1 | - | 7, 8 | 894 | |||
| Y18565.1 | - | 1, 2 | 40 | |||
| Y18566.1 | - | 5, 6 | 647 | |||
| Y18571.1 | - | 2, 3 | 122 | |||
| Y18572.1 | - | 7, 8 | 952 | |||

Masp2
Genomic Map