siRNA Details
Gene Symbol: Naa35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133324.1 | NP_579858.1 | 17 | 1587 | ||
GenBank mRNA | AF272892.1 | - | 17 | 1587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Naa35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133324.1 | NP_579858.1 | 17 | 1587 | ||
GenBank mRNA | AF272892.1 | - | 17 | 1587 |