siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133324.1 | NP_579858.1 | 12, 13 | 1109 | ||
| XM_006253526.2 | XP_006253588.1 | 12, 13 | 1374 | |||
| XM_006253527.1 | XP_006253589.1 | 10, 11 | 973 | |||
| GenBank mRNA | AF272892.1 | - | 12, 13 | 1109 | ||
| BC074005.1 | - | 12, 13 | 1086 | 7099269 | ||

Naa35
Genomic Map