siRNA Details
| Gene Symbol: Bpnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171990.2 | NP_741987.1 | 5, 6 | 462 | ||
| XM_006250429.2 | XP_006250491.1 | 5, 6 | 504 | |||
| XM_006250430.2 | XP_006250492.1 | 5, 6 | 559 | |||
| XM_006250431.2 | XP_006250493.1 | 5, 6 | 558 | |||
| GenBank mRNA | AJ000347.1 | - | 5, 6 | 413 | ||
| BC085692.1 | - | 5, 6 | 462 | 7190529 | ||
| FQ214355.1 | - | 5, 6 | 485 | |||

Bpnt1
Genomic Map