siRNA Details
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172062.2 | NP_742059.2 | 2, 3 | 321 | ||
XM_006256424.2 | XP_006256486.1 | 3, 4 | 406 | |||
XM_006256425.2 | XP_006256487.1 | 2, 3 | 315 | |||
GenBank mRNA | BC078703.1 | - | 2, 3 | 321 | 7115543 | |
X78949.1 | - | 2, 3 | 155 |
