siRNA Details
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172062.2 | NP_742059.2 | 9 | 1366 | ||
XM_006256424.2 | XP_006256486.1 | 10 | 1451 | |||
GenBank mRNA | BC078703.1 | - | 9 | 1366 | 7115543 | |
X78949.1 | - | 9 | 1200 |
