siRNA Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152861.4 | NP_690600.1 | 18 | 2243 | ||
| GenBank mRNA | D83948.1 | - | 18 | 2243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm10| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152861.4 | NP_690600.1 | 18 | 2243 | ||
| GenBank mRNA | D83948.1 | - | 18 | 2243 | ||