siRNA Details
| Gene Symbol: Pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033485.2 | NP_277020.1 | 5 | 916 | ||
| XM_006241317.2 | XP_006241379.1 | 6 | 1129 | |||
| GenBank mRNA | U05989.1 | - | 5 | 916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pawr| Gene Symbol: Pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033485.2 | NP_277020.1 | 5 | 916 | ||
| XM_006241317.2 | XP_006241379.1 | 6 | 1129 | |||
| GenBank mRNA | U05989.1 | - | 5 | 916 | ||