siRNA Details
| Gene Symbol: Cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171993.1 | NP_741990.1 | 2 | 444 | ||
| GenBank mRNA | BC085691.1 | - | 2 | 428 | 7134739 | |
| U05341.1 | - | 2 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc20| Gene Symbol: Cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171993.1 | NP_741990.1 | 2 | 444 | ||
| GenBank mRNA | BC085691.1 | - | 2 | 428 | 7134739 | |
| U05341.1 | - | 2 | 444 | |||