siRNA Details
| Gene Symbol: Mta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022588.1 | NP_072110.1 | 4 | 375 | ||
| XM_008764949.1 | XP_008763171.1 | 4 | 375 | |||
| XM_008764950.1 | XP_008763172.1 | 4 | 375 | |||
| XM_008764951.1 | XP_008763173.1 | 4 | 374 | |||
| XM_008764952.1 | XP_008763174.1 | 3 | 324 | |||
| XM_008764953.1 | XP_008763175.1 | 4 | 374 | |||
| XM_008764954.1 | XP_008763176.1 | 4 | 374 | |||
| XM_008764955.1 | XP_008763177.1 | 3 | 323 | |||
| XM_008764956.1 | XP_008763178.1 | 3 | 323 | |||
| XM_008764957.1 | XP_008763179.1 | 3 | 323 | |||
| XM_008764958.1 | XP_008763180.1 | 4 | 375 | |||
| GenBank mRNA | U02522.1 | - | 4 | 375 | ||

Mta1
Genomic Map