siRNA Details
| Gene Symbol: Mta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022588.1 | NP_072110.1 | 19 | 2074 | ||
| XM_008764949.1 | XP_008763171.1 | 21 | 2170 | |||
| XM_008764950.1 | XP_008763172.1 | 20 | 2158 | |||
| XM_008764951.1 | XP_008763173.1 | 21 | 2121 | |||
| XM_008764952.1 | XP_008763174.1 | 20 | 2119 | |||
| XM_008764953.1 | XP_008763175.1 | 20 | 2109 | |||
| XM_008764954.1 | XP_008763176.1 | 19 | 2073 | |||
| XM_008764955.1 | XP_008763177.1 | 20 | 2070 | |||
| XM_008764956.1 | XP_008763178.1 | 19 | 2058 | |||
| XM_008764957.1 | XP_008763179.1 | 18 | 2022 | |||
| GenBank mRNA | AJ132046.1 | - | 7 | 632 | ||
| U02522.1 | - | 19 | 2074 | |||

Mta1
Genomic Map