siRNA Details
| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022590.2 | NP_072112.2 | 11 | 1413 | ||
| GenBank mRNA | AB070849.1 | - | 10 | 1548 | ||
| AB070850.1 | - | 10 | 1548 | |||
| AB070976.1 | - | 11 | 1272 | |||
| U29881.1 | - | 11 | 1413 | |||

Slc5a2
Genomic Map