siRNA Details
| Gene Symbol: Ech1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022594.1 | NP_072116.1 | 2, 3 | 267 | ||
| GenBank mRNA | BC062226.1 | - | 2, 3 | 273 | 6890529 | |
| FQ214121.1 | - | 2, 3 | 269 | |||
| FQ230435.1 | - | 2, 3 | 280 | |||
| U08976.1 | - | 2, 3 | 267 | |||

Ech1
Genomic Map