siRNA Details
| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022610.1 | NP_072132.1 | 4, 5 | 622 | ||
| XM_006245038.2 | XP_006245100.1 | 6, 7 | 871 | |||
| GenBank mRNA | AB023134.1 | - | 4, 5 | 622 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icos| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022610.1 | NP_072132.1 | 4, 5 | 622 | ||
| XM_006245038.2 | XP_006245100.1 | 6, 7 | 871 | |||
| GenBank mRNA | AB023134.1 | - | 4, 5 | 622 | ||