siRNA Details
| Gene Symbol: Il12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022611.1 | NP_072133.1 | 6 | 899 | ||
| XM_006246147.2 | XP_006246209.1 | 7 | 1108 | |||
| GenBank mRNA | AF133197.1 | - | 6 | 899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12b| Gene Symbol: Il12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022611.1 | NP_072133.1 | 6 | 899 | ||
| XM_006246147.2 | XP_006246209.1 | 7 | 1108 | |||
| GenBank mRNA | AF133197.1 | - | 6 | 899 | ||