siRNA Details
| Gene Symbol: Mpeg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022617.1 | NP_072139.1 | 1 | 1321 | ||
| XM_006231115.2 | XP_006231177.1 | 2 | 1398 | |||
| XM_008760261.1 | XP_008758483.1 | 2 | 1490 | |||
| GenBank mRNA | AF156540.1 | - | 1 | 1321 | ||

Mpeg1
Genomic Map