siRNA Details
| Gene Symbol: Akap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022618.1 | NP_072140.1 | 13 | 5871 | ||
| GenBank mRNA | AF139518.1 | - | 13 | 5871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap6| Gene Symbol: Akap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022618.1 | NP_072140.1 | 13 | 5871 | ||
| GenBank mRNA | AF139518.1 | - | 13 | 5871 | ||