siRNA Details
| Gene Symbol: Bard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022622.1 | NP_072144.1 | 3 | 320 | ||
| GenBank mRNA | AF182946.1 | - | 3 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bard1| Gene Symbol: Bard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022622.1 | NP_072144.1 | 3 | 320 | ||
| GenBank mRNA | AF182946.1 | - | 3 | 320 | ||