siRNA Details
| Gene Symbol: Nphs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022628.1 | NP_072150.1 | 3 | 761 | ||
| XM_008759186.1 | XP_008757408.1 | 3 | 403 | |||
| XM_008759187.1 | XP_008757409.1 | 3 | 743 | |||
| GenBank mRNA | AF125521.1 | - | 3 | 304 | ||
| AF161715.1 | - | 3 | 761 | |||
| AF172255.1 | - | 3 | 388 | |||

Nphs1
Genomic Map