siRNA Details
| Gene Symbol: Adnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022681.2 | NP_073172.2 | 4 | 3027 | ||
| GenBank mRNA | AF234680.1 | - | 1 | 2047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adnp| Gene Symbol: Adnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022681.2 | NP_073172.2 | 4 | 3027 | ||
| GenBank mRNA | AF234680.1 | - | 1 | 2047 | ||