siRNA Details
| Gene Symbol: Cul5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022683.1 | NP_073174.1 | 9 | 1098 | ||
| XM_006243090.2 | XP_006243152.1 | 8 | 1811 | |||
| GenBank mRNA | AF135115.1 | - | 9 | 1098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul5| Gene Symbol: Cul5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022683.1 | NP_073174.1 | 9 | 1098 | ||
| XM_006243090.2 | XP_006243152.1 | 8 | 1811 | |||
| GenBank mRNA | AF135115.1 | - | 9 | 1098 | ||