siRNA Details
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022683.1 | NP_073174.1 | 10 | 1160 | ||
XM_006243090.2 | XP_006243152.1 | 9 | 1873 | |||
GenBank mRNA | AF135115.1 | - | 10 | 1160 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022683.1 | NP_073174.1 | 10 | 1160 | ||
XM_006243090.2 | XP_006243152.1 | 9 | 1873 | |||
GenBank mRNA | AF135115.1 | - | 10 | 1160 |