siRNA Details
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022694.2 | NP_073185.2 | 5, 6 | 781 | ||
| GenBank mRNA | AY697864.1 | - | 5, 6 | 791 | ||
| BC072471.1 | - | 5, 6 | 781 | 7100501 | ||
| U83883.1 | - | 5, 6 | 525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snd1| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022694.2 | NP_073185.2 | 5, 6 | 781 | ||
| GenBank mRNA | AY697864.1 | - | 5, 6 | 791 | ||
| BC072471.1 | - | 5, 6 | 781 | 7100501 | ||
| U83883.1 | - | 5, 6 | 525 | |||