siRNA Details
| Gene Symbol: Taok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022702.1 | NP_073193.1 | 14 | 1852 | ||
| XM_006230329.2 | XP_006230391.1 | 14 | 3293 | |||
| XM_006230330.2 | XP_006230392.1 | 13 | 1714 | |||
| XM_006230331.2 | XP_006230393.1 | 14 | 2581 | |||
| XM_006230332.2 | XP_006230394.1 | 14 | 2564 | |||
| XM_006230334.2 | XP_006230396.1 | 14 | 2566 | |||
| GenBank mRNA | AB290408.1 | - | 13 | 1678 | ||
| AF140556.2 | - | 14 | 1852 | |||
| FQ223093.1 | - | 14 | 2489 | |||

Taok2
Genomic Map