siRNA Details
| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022711.4 | NP_073202.1 | 2, 3 | 497 | ||
| GenBank mRNA | M95058.1 | - | 2, 3 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srd5a2| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022711.4 | NP_073202.1 | 2, 3 | 497 | ||
| GenBank mRNA | M95058.1 | - | 2, 3 | 497 | ||