siRNA Details
| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022713.2 | NP_073204.2 | 4 | 431 | ||
| XM_006244202.2 | XP_006244264.1 | 4 | 611 | |||
| GenBank mRNA | BC066665.1 | - | 4 | 420 | 5623927 | |
| DN934649.1 | - | 4 | 462 | |||
| M32725.1 | - | 1 | 72 | |||

Tgm4
Genomic Map