siRNA Details
| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031698.1 | NP_113886.1 | 16 | 1848 | ||
| XM_006235379.1 | XP_006235441.1 | 16 | 1880 | |||
| GenBank mRNA | BC060556.1 | - | 17 | 1858 | 6919638 | |
| X55298.1 | - | 16 | 1848 | |||

Rpn2
Genomic Map