siRNA Details
| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022800.1 | NP_073637.1 | 2 | 1056 | ||
| XM_006232406.2 | XP_006232468.1 | 3 | 1342 | |||
| XM_006232407.2 | XP_006232469.1 | 3 | 1432 | |||
| XM_006232408.2 | XP_006232470.1 | 3 | 2578 | |||
| GenBank mRNA | AF313450.1 | - | 2 | 1056 | ||
More Information
Set Membership: |
|

P2ry12
Genomic Map