siRNA Details
| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244902.2 | XP_006244964.1 | 2 | 477 | ||
| GenBank mRNA | FM034218.1 | - | 2 | 445 | ||
| FQ222344.1 | - | 3 | 453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nab1| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244902.2 | XP_006244964.1 | 2 | 477 | ||
| GenBank mRNA | FM034218.1 | - | 2 | 445 | ||
| FQ222344.1 | - | 3 | 453 | |||