siRNA Details
| Gene Symbol: N5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022857.1 | NP_074048.1 | 2 | 1231 | ||
| GenBank mRNA | L31882.1 | - | 2 | 1231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

N5| Gene Symbol: N5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022857.1 | NP_074048.1 | 2 | 1231 | ||
| GenBank mRNA | L31882.1 | - | 2 | 1231 | ||