siRNA Details
| Gene Symbol: B4galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022860.2 | NP_074051.1 | 5, 6 | 679 | ||
| XM_006241497.2 | XP_006241559.1 | 5, 6 | 1206 | |||
| XM_008765419.1 | XP_008763641.1 | 5, 6 | 3035 | |||
| XM_008765420.1 | XP_008763642.1 | 4, 5 | 1356 | |||
| GenBank mRNA | BC081799.1 | - | 5, 6 | 679 | 7133837 | |
| D17809.1 | - | 5, 6 | 568 | |||

B4galnt1
Genomic Map