siRNA Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022863.2 | NP_074054.2 | 6 | 787 | ||
| XM_006243091.2 | XP_006243153.1 | 6 | 1092 | |||
| XM_006243093.2 | XP_006243155.1 | 6 | 767 | |||
| GenBank mRNA | BC081798.1 | - | 6 | 787 | 7130997 | |
| U20181.1 | - | 6 | 692 | |||

Ireb2
Genomic Map