siRNA Details
| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022865.3 | NP_074056.2 | 14 | 1653 | ||
| GenBank mRNA | EU735541.1 | - | 15 | 1568 | ||
| X66366.2 | - | 14 | 1783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gphn| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022865.3 | NP_074056.2 | 14 | 1653 | ||
| GenBank mRNA | EU735541.1 | - | 15 | 1568 | ||
| X66366.2 | - | 14 | 1783 | |||