siRNA Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022866.2 | NP_074057.2 | 8 | 1099 | ||
| GenBank mRNA | AF080451.1 | - | 8 | 1076 | ||
| AF081825.1 | - | 8 | 1101 | |||
| BC129094.1 | - | 8 | 1099 | 8365494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a3| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022866.2 | NP_074057.2 | 8 | 1099 | ||
| GenBank mRNA | AF080451.1 | - | 8 | 1076 | ||
| AF081825.1 | - | 8 | 1101 | |||
| BC129094.1 | - | 8 | 1099 | 8365494 | ||